accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHCH3

CAS RN 2470-68-0

IUPAC Name: diheptylamine
Synonyms: 1-heptanamine, N-heptyl-
Molecular Weight (g/mol): 213.41
Molecular Formula: C14H31N
InChi Key: NJWMENBYMFZACG-UHFFFAOYSA-N
SMILES: CCCCCCCNCCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results