accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONNN

CAS RN 247058-06-6

IUPAC Name: methyl 6-(pyridin-2-yl)-[2,2'-bipyridine]-4-carboxylate
Synonyms: methyl 6-(pyridin-2-yl)-[2,2'-bipyridine]-4-carboxylate
Molecular Weight (g/mol): 291.31
Molecular Formula: C17H13N3O2
InChi Key: GEXSUZVSFVGSMZ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results