CAS RN 247069-93-8
CAS RN 247069-93-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ ALDH Detection Reagent - BAAA-DA
Fluorescent ALDH substrate and detection reagent. Used for: identification of cells with high ALDH activity. Application: flow cytometry, fluorescent-activated cell sorting (FACS)