Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ALDH Detection Reagent - BAAA-DA
SDP

Catalog No. 7556100U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-561-00U 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 75-561-00U Supplier Tocris Bioscience™ Supplier No. 7556/100U
Add to Cart
Add to Cart
This item is not returnable. View return policy

Fluorescent ALDH substrate and detection reagent. Used for: identification of cells with high ALDH activity. Application: flow cytometry, fluorescent-activated cell sorting (FACS)

Key information: ALDH Detection Reagent - BAAA-DA is a fluorescent substrate of aldehyde dehydrogenase (ALDH).

Used for: can be used in flow cytometry to sort cancer stem cells with high ALDH levels, and to isolate human hematopoietic progenitor cells. Can be used with efflux inhibitors such as Verapamil (Cat. No. 0654), Probenecid (Cat. No. 4107) or 2-deoxy-D-glucose (Cat. No. 4515) to block multi-drug resistance efflux.

Application: flow cytometry, fluorescent-activated cell sorting (FACS).

Properties and Photophysical Data: BAAA-DA is converted under acidic conditions to BAAA. BAAA, cell-permeant, is converted by intracellular ALDH into carboxylate ion amino acetate (BAA) and used to identify cells with high ALDH activity. Excitation and emission maxima (λ) are 505 nm and 512 nm, respectively; extinction coefficient = 74,595 M-1cm-1.

Specifications

Chemical Name or Material (T-4)-[N-(2,2-Diethoxyethyl)-5-[(3,5-dimethyl-2H-pyrrol-2-ylidene-κN)methyl]-1H-pyrrole-2-propanamidato-κN1]difluoroboron
CAS 247069-93-8
Quantity 100 μg
Target Enzyme Probes and Enzyme Substrates
Molecular Formula C20H28BF2N3O3
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.