accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(R)(S)NCH3ONHCH3Cl(R)O(R)SH3C(R)OPHOHOO(S)OH(R)OH

CAS RN 24729-96-2

IUPAC Name: {[(2R,3R,4S,5R,6R)-6-(2-chloro-1-{[(2S,4R)-1-methyl-4-propylpyrrolidin-2-yl]formamido}propyl)-4,5-dihydroxy-2-(methylsulfanyl)oxan-3-yl]oxy}phosphonic acid
Synonyms: [(2R,3R,4S,5R,6R)-6-(2-chloro-1-{[(2S,4R)-1-methyl-4-propylpyrrolidin-2-yl]formamido}propyl)-4,5-dihydroxy-2-(methylsulfanyl)oxan-3-yl]oxyphosphonic acid
Molecular Weight (g/mol): 504.96
Molecular Formula: C18H34ClN2O8PS
InChi Key: UFUVLHLTWXBHGZ-KUWMELJBSA-N
SMILES: CCC[C@@H]1C[C@H](N(C)C1)C(=O)NC(C(C)Cl)[C@H]1O[C@H](SC)[C@H](OP(O)(O)=O)[C@@H](O)[C@H]1O
Color 
  • (1)
  • (5)
  • (2)
Physical Form 
  • (7)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Type 
  • (5)
  • (3)
Grade 
  • (2)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (2)
  • (1)
  • (5)
  • (1)
  • (5)
  • (2)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (5)
  • (3)
  • (2)
  • (5)
  • (5)
  • (2)