Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clindamycin Phosphate USP , Medisca™
GREENER_CHOICESDP

Catalog No. 18048129 Shop All Medisca Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-048-129 100 g
18-048-023 10 g
18-048-061 1 kg
18-048-105 25 g
18-048-122 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-048-129 Supplier Medisca Supplier No. 000605
Get 1 Extra Chemical When You Buy 4
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Clindamycin Phosphate USP is a white to off-white, hygroscopic, crystalline powder. It is odorless to practically odorless.

Chemical Identifiers

CAS 24729-96-2

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Crystalline Powder
Color White to Off-White
Quantity 100 g
Percent Purity ≥ 758 μg/mg
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.