accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NH2ONH2NH2O

CAS RN 2475-45-8

IUPAC Name: 1,4,5,8-tetraamino-9,10-dihydroanthracene-9,10-dione
Synonyms: 1,4,5,8-tetraaminoanthracene-9,10-dione
Molecular Weight (g/mol): 268.28
Molecular Formula: C14H12N4O2
InChi Key: JSFUMBWFPQSADC-UHFFFAOYSA-N
SMILES: NC1=CC=C(N)C2=C1C(=O)C1=C(N)C=CC(N)=C1C2=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results