CAS RN 247580-43-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ SB 328437
A highly potent and selective CC chemokine receptor-3 (CCR3) antagonist.