accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOCH3H2N

CAS RN 24812-90-6

IUPAC Name: methyl 3-amino-4-methoxybenzoate
Synonyms: methyl 3-amino-4-methoxybenzoate
Molecular Weight (g/mol): 181.19
Molecular Formula: C9H11NO3
InChi Key: QVDWKLDUBSJEOG-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(OC)C(N)=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results