accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OOHNHNOHOO

CAS RN 2483-47-8

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-6-formamidohexanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-6-formamidohexanoic acid
Molecular Weight (g/mol): 274.32
Molecular Formula: C12H22N2O5
InChi Key: QYCPNUMTVZBTMM-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CCCCNC=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)