accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2OH2OH2OH2OH2OH2ONaNaONOONNONOONOONNONO(E)(E)

CAS RN 24868-20-0

IUPAC Name: disodium bis(3-[(E)-{[5-(4-nitrophenyl)furan-2-yl]methylidene}amino]-2,5-dioxoimidazolidin-1-ide) heptahydrate
Synonyms: dantrolene sodium hemiheptahydrate
Molecular Weight (g/mol): 798.58
Molecular Formula: C28H32N8Na2O17
InChi Key: LTWQNYPDAUSXBC-CDJGKPBYSA-L
SMILES: O.O.O.O.O.O.O.[Na+].[Na+].[O-][N+](=O)C1=CC=C(C=C1)C1=CC=C(O1)\C=N\N1CC(=O)[N-]C1=O.[O-][N+](=O)C1=CC=C(C=C1)C1=CC=C(O1)\C=N\N1CC(=O)[N-]C1=O
Grade 
  • (4)
Percent Purity 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (4)
  • (2)
  • (4)
  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)