Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dantrolene sodium hemiheptahydrate, Thermo Scientific™

Catalog No. AC469940250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469940250 25 g
AC469940050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC469940250 Supplier Thermo Scientific Chemicals Supplier No. 469940250
Add to Cart
This item is not returnable. View return policy

Specifications

Chemical Name or Material Dantrolene sodium hemiheptahydrate
CAS 24868-20-0
Synonym 1-[(5-(p-Nitrophenyl)-furfurylidene)amino]hydantoin sodium salt
Quantity 25 g
Solubility slightly soluble aq. soln.
Melting Point 225°C to 226°C
Shipping Condition No special requirements
Product Line Life science
Molecular Formula C14 H9 N4 Na O5 · 3.5 H2O
Physical Form Crystalline Powder
Color Yellow to Orange to Brown
Purity ≥97.5%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.