accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHBrBr

CAS RN 24898-17-7

IUPAC Name: bis[(4-bromophenyl)methyl]amine
Synonyms: bis[(4-bromophenyl)methyl]amine
Molecular Weight (g/mol): 355.07
Molecular Formula: C14H13Br2N
InChi Key: DNHBOKSBUKGKMI-UHFFFAOYSA-N
SMILES: BrC1=CC=C(CNCC2=CC=C(Br)C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results