Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Bis(4-bromobenzyl)amine 98.0+%, TCI America™
SDP

Catalog No. B21915G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B21915G Supplier TCI America Supplier No. B21915G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 24898-17-7
Molecular Formula C14H13Br2N
Molecular Weight (g/mol) 355.073
MDL Number MFCD03093622
InChI Key DNHBOKSBUKGKMI-UHFFFAOYSA-N
PubChem CID 10601487
IUPAC Name 1-(4-bromophenyl)-N-[(4-bromophenyl)methyl]methanamine
SMILES C1=CC(=CC=C1CNCC2=CC=C(C=C2)Br)Br

Specifications

Melting Point 51°C
Color White-Yellow
Quantity 5 g
Formula Weight 355.07
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Bis(4-bromobenzyl)amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.