accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPSOH3CSCl

CAS RN 24934-91-6

IUPAC Name: O,O-diethyl [(chloromethyl)sulfanyl]phosphonothioate
Synonyms: dotan C5H12ClO2PS2 caswell no. 195B epa pesticide chemical code 295300 chlormefos chlormephos chlormethylfos
Molecular Weight (g/mol): 234.69
Molecular Formula: C5H12ClO2PS2
InChi Key: QGTYWWGEWOBMAK-UHFFFAOYSA-N
SMILES: CCOP(=S)(OCC)SCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results