accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2496-90-4

IUPAC Name: 2-hydroxyethyl 4-hydroxybenzoate
Synonyms: 2-hydroxyethyl 4-hydroxybenzoate
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: GFHGEIJFEHZKHZ-UHFFFAOYSA-N
SMILES: OCCOC(=O)C1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results