Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxyethyl 4-Hydroxybenzoate 98.0+%, TCI America™
SDP

Catalog No. H069810G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H069810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H069810G Supplier TCI America Supplier No. H069810G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2496-90-4
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00059079
InChI Key GFHGEIJFEHZKHZ-UHFFFAOYSA-N
Synonym 4-Hydroxybenzoic Acid 2-Hydroxyethyl Ester, 2-Hydroxyethylparaben
PubChem CID 267950
IUPAC Name 2-hydroxyethyl 4-hydroxybenzoate
SMILES OCCOC(=O)C1=CC=C(O)C=C1

Specifications

Melting Point 141°C
Color White
Boiling Point 202°C
Quantity 10 g
Formula Weight 182.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxyethyl 4-Hydroxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.