CAS RN 25013-16-5
Butylated hydroxyanisole, 96%
CAS: 25013-16-5 Molecular Formula: C11H16O2 Molecular Weight (g/mol): 180.24 MDL Number: MFCD01779059 InChI Key: MRBKEAMVRSLQPH-UHFFFAOYSA-N Synonym: 3-tert-butyl-4-hydroxyanisole,4-hydroxy-3-tert-butylanisole,2-tert-butyl-4-methoxyphenol,3-bha,4-methoxy-2-tert-butylphenol,3-t-butyl-4-hydroxyanisole,phenol, 2-1,1-dimethylethyl-4-methoxy,2-1,1-dimethylethyl-4-methoxyphenol,2-butyl-4-hydroxyanisole,phenol, 2-tert-butyl-4-methoxy PubChem CID: 8456 ChEBI: CHEBI:76358 IUPAC Name: 2-tert-butyl-4-methoxyphenol SMILES: CC(C)(C)C1=C(C=CC(=C1)OC)O
BHA, FCC, 98.5%, Spectrum™ Chemical
CAS: 25013-16-5 Molecular Formula: C11H16O2 Molecular Weight (g/mol): 180.25 InChI Key: MRBKEAMVRSLQPH-UHFFFAOYSA-N IUPAC Name: 2-tert-butyl-4-methoxyphenol SMILES: COC1=CC=C(O)C(=C1)C(C)(C)C
2(3)-tert-Butyl-4-methoxyphenol, 96%
CAS: 25013-16-5 Molecular Formula: C11H16O2 Molecular Weight (g/mol): 180.247 MDL Number: MFCD01779059 InChI Key: MRBKEAMVRSLQPH-UHFFFAOYSA-N Synonym: 3-tert-butyl-4-hydroxyanisole,4-hydroxy-3-tert-butylanisole,2-tert-butyl-4-methoxyphenol,3-bha,4-methoxy-2-tert-butylphenol,3-t-butyl-4-hydroxyanisole,phenol, 2-1,1-dimethylethyl-4-methoxy,2-1,1-dimethylethyl-4-methoxyphenol,2-butyl-4-hydroxyanisole,phenol, 2-tert-butyl-4-methoxy PubChem CID: 8456 ChEBI: CHEBI:76358 IUPAC Name: 2-tert-butyl-4-methoxyphenol SMILES: CC(C)(C)C1=C(C=CC(=C1)OC)O
Butylated hydroxyanisole Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Butylated Hydroxyanisole, 98.5%, MP Biomedicals™
CAS: 25013-16-5 Molecular Formula: C11H16O2 Molecular Weight (g/mol): 180.247 InChI Key: MRBKEAMVRSLQPH-UHFFFAOYSA-N Synonym: 3-tert-butyl-4-hydroxyanisole,4-hydroxy-3-tert-butylanisole,2-tert-butyl-4-methoxyphenol,3-bha,4-methoxy-2-tert-butylphenol,3-t-butyl-4-hydroxyanisole,phenol, 2-1,1-dimethylethyl-4-methoxy,2-1,1-dimethylethyl-4-methoxyphenol,2-butyl-4-hydroxyanisole,phenol, 2-tert-butyl-4-methoxy PubChem CID: 8456 ChEBI: CHEBI:76358 IUPAC Name: 2-tert-butyl-4-methoxyphenol SMILES: CC(C)(C)C1=C(C=CC(=C1)OC)O
Sigma Aldrich Butylated hydroxyanisole
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Butylated Hydroxyanisole, SPEX CertiPrep™
CAS: 25013-16-5 Molecular Formula: C11H16O2 Molecular Weight (g/mol): 180.247 InChI Key: MRBKEAMVRSLQPH-UHFFFAOYSA-N PubChem CID: 8456 ChEBI: CHEBI:76358 IUPAC Name: 2-tert-butyl-4-methoxyphenol SMILES: CC(C)(C)C1=C(C=CC(=C1)OC)O