accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25024-53-7

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-[1-(2,4-dinitrophenyl)-1H-imidazol-4-yl]propanoic acid
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3-[1-(2,4-dinitrophenyl)imidazol-4-yl]propanoic acid
Molecular Weight (g/mol): 421.37
Molecular Formula: C17H19N5O8
InChi Key: KPGVUOQMOHGHEW-LBPRGKRZSA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CC1=CN(C=N1)C1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(O)=O
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)