accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHSOONOONOONOO

CAS RN 2508-19-2

IUPAC Name: 2,4,6-trinitrobenzene-1-sulfonic acid
Synonyms: trinitrobenzenesulfonic acid TNBS
Molecular Weight (g/mol): 293.16
Molecular Formula: C6H3N3O9S
InChi Key: NHJVRSWLHSJWIN-UHFFFAOYSA-N
SMILES: OS(=O)(=O)C1=C(C=C(C=C1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)