accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONNH3CNOOOHN(E/Z)

CAS RN 2512-29-0

IUPAC Name: 2-[2-(4-methyl-2-nitrophenyl)diazen-1-yl]-3-oxo-N-phenylbutanamide
Synonyms: 2-[2-(4-methyl-2-nitrophenyl)diazen-1-yl]-3-oxo-N-phenylbutanamide
Molecular Weight (g/mol): 340.34
Molecular Formula: C17H16N4O4
InChi Key: MFYSUUPKMDJYPF-UHFFFAOYNA-N
SMILES: CC(=O)C(N=NC1=CC=C(C)C=C1[N+]([O-])=O)C(=O)NC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results