accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 251456-60-7

IUPAC Name: 7-{[4-(dimethylamino)phenyl]formamido}-N-hydroxyheptanamide
Synonyms: 7-{[4-(dimethylamino)phenyl]formamido}-N-hydroxyheptanamide
Molecular Weight (g/mol): 307.39
Molecular Formula: C16H25N3O3
InChi Key: MXWDSZWTBOCWBK-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)C(=O)NCCCCCCC(=O)NO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results