Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Dimethylamino)-N-[7-(hydroxyamino)-7-oxoheptyl]benzamide 97.0+%, TCI America™
SDP

Catalog No. D418820MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D418820MG 20 mg
D4188100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D418820MG Supplier TCI America Supplier No. D418820MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 251456-60-7
Molecular Formula C16H25N3O3
Molecular Weight (g/mol) 307.394
MDL Number MFCD03453554
InChI Key MXWDSZWTBOCWBK-UHFFFAOYSA-N
Synonym 4-dimethylamino-n-7-hydroxyamino-7-oxoheptyl benzamide, histone deacetylase inhibitor iii, n-hydroxy-7-4-dimethylaminobenzoyl aminoheptanamide, 4-dimethylamino-n-7-hydroxyamino-7-oxoheptyl-benzamide, 4-dimethylamino-n-6-hydroxycarbamoylhexyl benzamide, n-hydroxy-7-4-dimethylaminobenzoyl-aminoheptanamide, 4-dimethylamino-n-6-hydroxycarbamoylhexyl-benzamide, benzamide, 4-dimethylamino-n-7-hydroxyamino-7-oxoheptyl, 7-4-dimethylamino phenyl formamido-n-hydroxyheptanamide
PubChem CID 3994
IUPAC Name 4-(dimethylamino)-N-[7-(hydroxyamino)-7-oxoheptyl]benzamide
SMILES CN(C)C1=CC=C(C=C1)C(=O)NCCCCCCC(=O)NO

Specifications

Melting Point 152°C
Color Yellow
Quantity 20 mg
Formula Weight 307.39
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Dimethylamino)-N-[7-(hydroxyamino)-7-oxoheptyl]benzamide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.