accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO(S)OOSH3COOOHO

CAS RN 251565-85-2

IUPAC Name: (2S)-2-ethoxy-3-(4-{2-[4-(methanesulfonyloxy)phenyl]ethoxy}phenyl)propanoic acid
Synonyms: galida tesaglitazar
Molecular Weight (g/mol): 408.47
Molecular Formula: C20H24O7S
InChi Key: CXGTZJYQWSUFET-IBGZPJMESA-N
SMILES: CCO[C@@H](CC1=CC=C(OCCC2=CC=C(OS(C)(=O)=O)C=C2)C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)