accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3CH3CCH3CH3CH3CH3CCH3CH3CH3CH3CCH3Ce

CAS RN 251984-08-4

IUPAC Name: cerium(3+) tris(1,2,3,4-tetramethylcyclopenta-2,4-dien-1-ide)
Synonyms: cerium(3+) tris(1,2,3,4-tetramethylcyclopenta-2,4-dien-1-ide)
Molecular Weight (g/mol): 503.73
Molecular Formula: C27H39Ce
InChi Key: VNYOSWSTNAVLMJ-UHFFFAOYSA-N
SMILES: [Ce+3].C[C-]1C=C(C)C(C)=C1C.C[C-]1C=C(C)C(C)=C1C.C[C-]1C=C(C)C(C)=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results