Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(tetramethylcyclopentadienyl)cerium(III), 98%, Thermo Scientific™

Catalog No. AA4438003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4438003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4438003 Supplier Thermo Scientific Chemicals Supplier No. 4438003

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 251984-08-4
Molecular Formula C27H39Ce
Molecular Weight (g/mol) 503.725
MDL Number MFCD01863662
InChI Key VNYOSWSTNAVLMJ-UHFFFAOYSA-N
Synonym tris tetramethylcyclopentadienyl cerium iii, tris 1,2,3,4-tetramethyl-2,4-cyclopentadienyl cerium iii
PubChem CID 121233288
IUPAC Name cerium(3+);1,2,3,5-tetramethylcyclopenta-1,3-diene
SMILES C[C-]1C=C(C(=C1C)C)C.C[C-]1C=C(C(=C1C)C)C.C[C-]1C=C(C(=C1C)C)C.[Ce+3]

Specifications

Color Green
Quantity 1 g
UN Number UN3181
Sensitivity Moisture Sensitive
Formula Weight 503.73
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Tris (tetramethylcyclopentadienyl)cerium (III)
Hazard Category Warning
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.