accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH2

CAS RN 2523-56-0

IUPAC Name: 4-methylcyclohexan-1-amine
Synonyms: 4-methylcyclohexan-1-amine
Molecular Weight (g/mol): 113.2
Molecular Formula: C7H15N
InChi Key: KSMVBYPXNKCPAJ-UHFFFAOYSA-N
SMILES: CC1CCC(N)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results