accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25236-64-0

CH3SOOOFFF

CAS RN 25236-64-0

IUPAC Name: 2,2,2-trifluoroethyl methanesulfonate
Synonyms: 2,2,2-trifluoroethyl methanesulfonate
Molecular Weight (g/mol): 178.13
Molecular Formula: C3H5F3O3S
InChi Key: ICECLJDLAVVEOW-UHFFFAOYSA-N
SMILES: CS(=O)(=O)OCC(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results