accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClPOOO

CAS RN 2524-64-3

IUPAC Name: diphenyl phosphorochloridate
Synonyms: diphenyl phosphorochloridate
Molecular Weight (g/mol): 268.63
Molecular Formula: C12H10ClO3P
InChi Key: BHIIGRBMZRSDRI-UHFFFAOYSA-N
SMILES: ClP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1
Boiling Point 
  • (3)
  • (2)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (6)
Percent Purity 
  • (3)
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (3)
  • (6)
  • (3)
  • (2)
  • (3)
  • (3)
  • (5)
  • (6)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)