CAS RN 25281-63-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MP Biomedicals, Inc Poly-DL-alanine, 95%, MP Biomedicals™
CAS: 25281-63-4 Molecular Formula: C3H7NO2 MDL Number: MFCD00166369