Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly-DL-alanine, 95%, MP Biomedicals™

Catalog No. ICN10267783 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10267783 250 mg
ICN10267780 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10267783 Supplier MP Biomedicals, Inc Supplier No. 0210267783

Chemical Identifiers

CAS 25281-63-4
Molecular Formula C3H7NO2
MDL Number MFCD00166369

Specifications

Color White
Assay Percent Range 95%
Quantity 250 mg
Formula Weight 1000 to 5000
Physical Form Powder
Chemical Name or Material Poly-DL-alanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.