accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaH2NSOOO

CAS RN 25293-52-1

IUPAC Name: sodium 2-aminonaphthalene-1-sulfonate
Synonyms: sodium 2-aminonaphthalene-1-sulfonate
Molecular Weight (g/mol): 245.23
Molecular Formula: C10H8NNaO3S
InChi Key: AZKQWFQWHHSLDU-UHFFFAOYSA-M
SMILES: [Na+].NC1=CC=C2C=CC=CC2=C1S([O-])(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results