accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2544-31-2

IUPAC Name: 2-amino-3-{[(4-methoxyphenyl)methyl]sulfanyl}propanoic acid
Synonyms: 2-amino-3-{[(4-methoxyphenyl)methyl]sulfanyl}propanoic acid
Molecular Weight (g/mol): 241.31
Molecular Formula: C11H15NO3S
InChi Key: PQPZSPJVMUCVAQ-UHFFFAOYNA-N
SMILES: COC1=CC=C(CSCC(N)C(O)=O)C=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)