accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3NHSHN(S)OH

CAS RN 254753-54-3

IUPAC Name: ethyl (4S)-4-(3-hydroxyphenyl)-6-methyl-2-sulfanylidene-1,2,3,4-tetrahydropyrimidine-5-carboxylate
Synonyms: monastrol
Molecular Weight (g/mol): 292.35
Molecular Formula: C14H16N2O3S
InChi Key: LOBCDGHHHHGHFA-LBPRGKRZSA-N
SMILES: CCOC(=O)C1=C(C)NC(=S)N[C@H]1C1=CC=CC(O)=C1
Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results