accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOCH3CH3CH3CH3H3CH3CBOOH3CH3CH3CH3C

CAS RN 254755-24-3

IUPAC Name: 2-[9,9-dihexyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-fluoren-2-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-[9,9-dihexyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)fluoren-2-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 586.47
Molecular Formula: C37H56B2O4
InChi Key: SYMMYBWUPCWTEI-UHFFFAOYSA-N
SMILES: CCCCCCC1(CCCCCC)C2=C(C=CC(=C2)B2OC(C)(C)C(C)(C)O2)C2=C1C=C(C=C2)B1OC(C)(C)C(C)(C)O1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results