CAS RN 2550-06-3
IUPAC Name:
trichloro(3-chloropropyl)silane
Synonyms:
trichloro(3-chloropropyl)silane
Molecular Weight (g/mol):
211.97
Molecular Formula:
C3H6Cl4Si
InChi Key:
OOXSLJBUMMHDKW-UHFFFAOYSA-N
SMILES:
ClCCC[Si](Cl)(Cl)Cl