accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2553-71-1

IUPAC Name: 3,3-bis(3-bromo-5-chloro-4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Synonyms: 3,3-bis(3-bromo-5-chloro-4-hydroxyphenyl)-2,1λ⁶-benzoxathiole-1,1-dione
Molecular Weight (g/mol): 581.05
Molecular Formula: C19H10Br2Cl2O5S
InChi Key: MDGFKZKMIQQRPU-UHFFFAOYSA-N
SMILES: OC1=C(Cl)C=C(C=C1Br)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Cl)=C(O)C(Br)=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (4)
Grade 
  • (4)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (4)
  • (1)