Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromochlorophenol Blue, TCI America™
SDP

Catalog No. B05741G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B05741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B05741G Supplier TCI America Supplier No. B05741G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2553-71-1
Molecular Formula C19H10Br2Cl2O5S
Molecular Weight (g/mol) 581.052
MDL Number MFCD00047291
InChI Key MDGFKZKMIQQRPU-UHFFFAOYSA-N
PubChem CID 73120
IUPAC Name 2-bromo-4-[3-(3-bromo-5-chloro-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-6-chlorophenol
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC(=C(C(=C3)Br)O)Cl)C4=CC(=C(C(=C4)Br)O)Cl

Specifications

Quantity 1 g
Formula Weight 581.05
Physical Form Crystalline Powder
Chemical Name or Material Bromochlorophenol Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.