accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25550-52-1

IUPAC Name: 3,5-dimethylbenzene-1-thiol
Synonyms: 3,5-dimethylbenzenethiol
Molecular Weight (g/mol): 138.23
Molecular Formula: C8H10S
InChi Key: CESBAYSBPMVAEI-UHFFFAOYSA-N
SMILES: CC1=CC(S)=CC(C)=C1
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results