accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiCH3CH3ONSiH3CH3CCH3FFF(E/Z)

CAS RN 25561-30-2

IUPAC Name: trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)ethanimidate
Synonyms: bstfa
Molecular Weight (g/mol): 257.4
Molecular Formula: C8H18F3NOSi2
InChi Key: XCOBLONWWXQEBS-UHFFFAOYSA-N
SMILES: C[Si](C)(C)OC(=N[Si](C)(C)C)C(F)(F)F
Molecular Weight (g/mol) 
  • (5)
  • (1)
  • (3)
  • (3)
  • (17)
  • (21)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (6)
  • (2)
  • (7)
  • (4)
  • (1)
  • (7)
  • (6)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (5)
  • (12)
  • (3)
  • (17)
  • (3)
  • (1)
  • (5)
Physical Form 
  • (36)
Percent Purity 
  • (4)
  • (3)
  • (4)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (12)
  • (2)
  • (18)
  • (3)
  • (5)
  • (23)
  • (1)
  • (5)
  • (1)
  • (3)
  • (3)
  • (17)
  • (21)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (6)
  • (2)
  • (7)
  • (4)
  • (1)
  • (7)
  • (6)
  • (1)
  • (1)
  • (2)
  • (5)
  • (12)
  • (3)
  • (17)
  • (3)
  • (1)
  • (5)
  • (36)
  • (4)
  • (3)
  • (4)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (4)
  • (3)
  • (1)
  • (17)
  • (17)