accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25595-59-9

IUPAC Name: 3-chloro-3H-2,1λ⁶-benzoxathiole-1,1-dione
Synonyms: 3-chloro-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular Weight (g/mol): 204.62
Molecular Formula: C7H5ClO3S
InChi Key: LNWUNIOANJTGPG-UHFFFAOYNA-N
SMILES: ClC1OS(=O)(=O)C2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results