Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Chloro-α-hydroxy-2-toluenesulfonic Acidγ-sultone 99%, Thermo Scientific™

Catalog No. AC156530050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC156530050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC156530050 Supplier Thermo Scientific Chemicals Supplier No. 156530050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25595-59-9
Molecular Formula C7H5ClO3S
Molecular Weight (g/mol) 204.624
MDL Number MFCD00005867
InChI Key LNWUNIOANJTGPG-UHFFFAOYSA-N
Synonym 3-chloro-3h-2,1-benzoxathiole-1,1-dioxide, 3-chloro-3h-2,1-benzoxathiole1,1-dioxide, 3-chloro-3h-benzo c 1,2-oxathiolene-1,1-dione, 3-chloro-3h-2,1??-benzoxathiole-1,1-dione, alpha-chloro-alpha,2-toluenesultone, 3-chloro-3h-2,1, 3-chloro-3h-2,1-benzoxathiole 1,1-dioxide, 3-chloro-3h-2,1-benzoxathiole 1,1-dioxide #, 3h-2,1-benzoxathiole,3-chloro-, 1,1-dioxide, 3-chloro-3h-benzo c 1,2 oxathiole 1,1-dioxide
PubChem CID 98353
IUPAC Name 3-chloro-3H-2,1$l^{6}-benzoxathiole 1,1-dioxide
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 110°C to 116°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Quantity 5 g
Formula Weight 204.63
Percent Purity ≥98.5%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material α-Chloro-α-hydroxy-2-toluenesulfonic acid γ-sultone, 99%
Show More Show Less
Hazard Category MOISTURE SENSITIVE
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.