accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHOOCH3(Z)(E)

CAS RN 25637-84-7

IUPAC Name: 2-hydroxy-3-[(9Z)-octadec-9-enoyloxy]propyl (9E)-octadec-9-enoate
Synonyms: 2-hydroxy-3-[(9Z)-octadec-9-enoyloxy]propyl (9E)-octadec-9-enoate glyceryl dioleate
Molecular Weight (g/mol): 621
Molecular Formula: C39H72O5
InChi Key: DRAWQKGUORNASA-YAFCTCPENA-N
SMILES: CCCCCCCC\C=C\CCCCCCCC(=O)OCC(O)COC(=O)CCCCCCC\C=C/CCCCCCCC
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results