CAS RN 25646-77-9
IUPAC Name:
2-[(4-amino-3-methylphenyl)(ethyl)amino]ethan-1-ol; sulfuric acid
Synonyms:
CD 4 color developer; sulfuric acid
Molecular Weight (g/mol):
292.35
Molecular Formula:
C11H20N2O5S
InChi Key:
GVEYRUKUJCHJSR-UHFFFAOYSA-N
SMILES:
OS(O)(=O)=O.CCN(CCO)C1=CC=C(N)C(C)=C1