accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2568-90-3

IUPAC Name: 1-(butoxymethoxy)butane
Synonyms: dibutoxymethane
Molecular Weight (g/mol): 160.26
Molecular Formula: C9H20O2
InChi Key: QLCJOAMJPCOIDI-UHFFFAOYSA-N
SMILES: CCCCOCOCCCC
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)