Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibutoxymethane 98.0+%, TCI America™
SDP

Catalog No. D171125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D171125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D171125G Supplier TCI America Supplier No. D171125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2568-90-3
Molecular Formula C9H20O2
Molecular Weight (g/mol) 160.26
MDL Number MFCD00059270
InChI Key QLCJOAMJPCOIDI-UHFFFAOYSA-N
Synonym Formaldehyde Dibutyl Acetal, 5,7-Dioxahendecane
PubChem CID 17379
IUPAC Name 1-(butoxymethoxy)butane
SMILES CCCCOCOCCCC

Specifications

Color Colorless
Boiling Point 180°C
Quantity 25 g
UN Number 1993
Formula Weight 160.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dibutoxymethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.