CAS RN 25717-80-0
IUPAC Name:
5-[(ethoxycarbonyl)azanidyl]-3-(morpholin-4-yl)-3H-1λ⁴,2,3-oxadiazol-1-ylium
Synonyms:
molsidomine
Molecular Weight (g/mol):
242.24
Molecular Formula:
C9H14N4O4
InChi Key:
XLFWDASMENKTKL-UHFFFAOYSA-N
SMILES:
CCOC(=O)[N-]C1=CN(N=[O+]1)N1CCOCC1
1
–
1
of
1
results
1
Molsidomine 98.0+%, TCI America™
CAS: 25717-80-0 Molecular Formula: C9H14N4O4 Molecular Weight (g/mol): 242.235 MDL Number: MFCD00869301 InChI Key: XLFWDASMENKTKL-UHFFFAOYSA-N PubChem CID: 5353788 IUPAC Name: (1E)-1-ethoxy-N-(3-morpholin-4-yloxadiazol-3-ium-5-yl)methanimidate SMILES: CCOC(=NC1=C[N+](=NO1)N2CCOCC2)[O-]