Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molsidomine 98.0+%, TCI America™
SDP

Catalog No. M25245G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25245G 5 g
M25241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25245G Supplier TCI America Supplier No. M25245G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25717-80-0
Molecular Formula C9H14N4O4
Molecular Weight (g/mol) 242.235
MDL Number MFCD00869301
InChI Key XLFWDASMENKTKL-UHFFFAOYSA-N
PubChem CID 5353788
IUPAC Name (1E)-1-ethoxy-N-(3-morpholin-4-yloxadiazol-3-ium-5-yl)methanimidate
SMILES CCOC(=NC1=C[N+](=NO1)N2CCOCC2)[O-]

Specifications

Melting Point 141°C
Color White
Quantity 5 g
Formula Weight 242.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Molsidomine
EINECSNumber (9)-2224
RTECSNumber WU7677400
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.