accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25729-13-9

IUPAC Name: 4-(nonyloxy)-N-{[4-(nonyloxy)phenyl]imino}benzen-1-imine oxide
Synonyms: 4-(nonyloxy)-N-{[4-(nonyloxy)phenyl]imino}benzenimine oxide
Molecular Weight (g/mol): 482.71
Molecular Formula: C30H46N2O3
InChi Key: KJMHEFPNHRGGGG-UHFFFAOYSA-N
SMILES: CCCCCCCCCOC1=CC=C(C=C1)N=[N+]([O-])C1=CC=C(OCCCCCCCCC)C=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results