accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONHSOOOCH3OH3C(E)

CAS RN 257292-29-8

IUPAC Name: 3-[(5E)-7-(2,4-dimethoxyphenyl)-1,4-thiazepan-5-ylidene]-6-methyl-3,4-dihydro-2H-pyran-2,4-dione
Synonyms: 3-[(5E)-7-(2,4-dimethoxyphenyl)-1,4-thiazepan-5-ylidene]-6-methylpyran-2,4-dione
Molecular Weight (g/mol): 375.44
Molecular Formula: C19H21NO5S
InChi Key: FJDFKOSFDPIMBV-NBVRZTHBNA-N
SMILES: COC1=CC=C(C2C\C(NCCS2)=C2/C(=O)OC(C)=CC2=O)C(OC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results